Assay Details
| Gene Symbol: PKDCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206721.1 | NP_001193650.1 | 1 - 2 | 803 | 60 | |
| GenBank mRNA | AV607688.1 | - | 1 - 2 | 75 | 60 | |
More Information
Set Membership: |
|

PKDCC
Genomic Map