Assay Details
| Gene Symbol: DPY19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206782.1 | NP_001193711.1 | 20 - 21 | 2191 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPY19L2| Gene Symbol: DPY19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206782.1 | NP_001193711.1 | 20 - 21 | 2191 | 99 | |
Set Membership: |
|