Assay Details
| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206792.1 | NP_001193721.1 | 1 - 2 | 343 | 72 | |
| GenBank mRNA | BE751627.1 | - | 1 - 2 | 343 | 72 | |
| EH173604.1 | - | 1 - 2 | 317 | 72 | ||
More Information
Set Membership: |
|

NCR3LG1
Genomic Map