Assay Details
Gene Symbol: THBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166522.1 | NP_001159994.1 | 1 | 3421 | 90 | |
GenBank mRNA | BC149343.1 | - | 1 | 3427 | 90 | |
M14657.1 | - | 1 | 2643 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
