Assay Details
| Gene Symbol: ABRACL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166561.1 | NP_001160033.1 | 1 - 2 | 106 | 96 | |
| GenBank mRNA | BC103206.1 | - | 1 - 2 | 83 | 96 | |
| EV665794.1 | - | 1 - 2 | 112 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103206.1, NM_001166561.1 transcripts.
Set Membership: |
|

ABRACL
Genomic Map