Assay Details
| Gene Symbol: ABRACL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166561.1 | NP_001160033.1 | 2 - 3 | 177 | 70 | |
| GenBank mRNA | BC103206.1 | - | 2 - 3 | 154 | 70 | |
| EV665794.1 | - | 2 - 3 | 183 | 70 | ||
More Information
Set Membership: |
|

ABRACL
Genomic Map