Assay Details
| Gene Symbol: TMEM216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166563.2 | NP_001160035.1 | 2 - 3 | 159 | 113 | |
| GenBank mRNA | BC111187.1 | - | 2 - 3 | 160 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111187.1, NM_001166563.2 transcripts.
Set Membership: |
|

TMEM216
Genomic Map