Assay Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166564.1 | NP_001160036.1 | 3 - 4 | 552 | 66 | |
| GenBank mRNA | BC142525.1 | - | 3 - 4 | 528 | 66 | |
More Information
Set Membership: |
|

FAAP20
Genomic Map