Assay Details
| Gene Symbol: SCAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166565.1 | NP_001160037.1 | 3 - 4 | 312 | 66 | |
| GenBank mRNA | BC118386.1 | - | 3 - 4 | 314 | 66 | |
More Information
Set Membership: |
|

SCAMP5
Genomic Map