Assay Details
| Gene Symbol: TRPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034392.4 | NP_001029564.1 | 5 - 6 | 543 | 82 | |
| NM_001166602.3 | NP_001160074.1 | 6 - 7 | 630 | 82 | ||
| GenBank mRNA | BC103210.1 | - | 5 - 6 | 514 | 82 | |
More Information
Set Membership: |
|

TRPT1
Genomic Map