Assay Details
| Gene Symbol: HSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166608.1 | NP_001160080.1 | 2 - 3 | 530 | 70 | |
| NM_001166609.1 | NP_001160081.1 | 2 - 3 | 415 | 70 | ||
| NM_001166610.1 | NP_001160082.1 | 2 - 3 | 282 | 70 | ||
| GenBank mRNA | AY911359.1 | - | 2 - 3 | 259 | 70 | |
| BC102077.1 | - | 2 - 3 | 233 | 70 | ||
More Information
Set Membership: |
|

HSPD1
Genomic Map