Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166611.1 | NP_001160083.1 | 3 - 4 | 361 | 57 | |
GenBank mRNA | BC148137.1 | - | 3 - 4 | 310 | 57 | |
EV659412.1 | - | 3 - 4 | 370 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
