Assay Details
| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166614.1 | NP_001160086.1 | 4 - 5 | 549 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPH10B| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166614.1 | NP_001160086.1 | 4 - 5 | 549 | 78 | |
Set Membership: |
|