Assay Details
| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166614.1 | NP_001160086.1 | 9 - 10 | 1271 | 94 | |
| GenBank mRNA | BT025392.1 | - | 8 - 9 | 1220 | 94 | |
More Information
Set Membership: |
|

RSPH10B
Genomic Map