Assay Details
| Gene Symbol: NFATC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166615.2 | NP_001160087.1 | 8 - 9 | 2285 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFATC1| Gene Symbol: NFATC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166615.2 | NP_001160087.1 | 8 - 9 | 2285 | 61 | |
Set Membership: |
|