Assay Details
| Gene Symbol: ADSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167809.1 | NP_001161281.1 | 10 - 11 | 1167 | 102 | |
| GenBank mRNA | BC151541.1 | - | 10 - 11 | 1136 | 102 | |
More Information
Set Membership: |
|

ADSS2
Genomic Map