Assay Details
| Gene Symbol: ADSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167809.1 | NP_001161281.1 | 5 - 6 | 580 | 78 | |
| GenBank mRNA | BC151541.1 | - | 5 - 6 | 549 | 78 | |
| EV656093.1 | - | 5 - 6 | 590 | 78 | ||
More Information
Set Membership: |
|

ADSS2
Genomic Map