Assay Details
| Gene Symbol: GORAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167897.3 | NP_001161369.1 | 4 - 5 | 729 | 57 | |
| GenBank mRNA | BC142523.1 | - | 4 - 5 | 732 | 57 | |
More Information
Set Membership: |
|

GORAB
Genomic Map