Assay Details
| Gene Symbol: POLD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167904.3 | NP_001161376.1 | 4 - 5 | 384 | 62 | |
| GenBank mRNA | BC151727.1 | - | 4 - 5 | 294 | 62 | |
More Information
Set Membership: |
|

POLD3
Genomic Map