Assay Details
| Gene Symbol: KLF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168462.1 | NP_001161934.1 | 2 - 3 | 398 | 105 | |
| GenBank mRNA | AB530320.1 | - | 2 - 3 | 398 | 105 | |
| BC149823.1 | - | 2 - 3 | 323 | 105 | ||
More Information
Set Membership: |
|

KLF10
Genomic Map