Assay Details
| Gene Symbol: MR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190298.1 | NP_001177227.1 | 3 - 4 | 593 | 106 | |
| GenBank mRNA | EU792881.1 | - | 3 - 4 | 587 | 106 | |
| FJ028657.1 | - | 3 - 4 | 593 | 106 | ||
| FJ423041.1 | - | 2 - 3 | 359 | 106 | ||
More Information
Set Membership: |
|

MR1
Genomic Map