Assay Details
Gene Symbol: IRAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190299.2 | NP_001177228.1 | 7 - 8 | 755 | 69 | |
GenBank mRNA | EU528622.1 | - | 5 - 6 | 612 | 69 | |
EU528623.1 | - | 6 - 7 | 677 | 69 | ||
EU528624.1 | - | 7 - 8 | 829 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
