Assay Details
| Gene Symbol: LONRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191134.1 | NP_001178063.1 | 10 - 11 | 2124 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LONRF3| Gene Symbol: LONRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191134.1 | NP_001178063.1 | 10 - 11 | 2124 | 55 | |
Set Membership: |
|