Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191156.2 | NP_001178085.1 | 1 - 2 | 267 | 101 | |
| GenBank mRNA | BC120411.1 | - | 1 - 2 | 267 | 101 | |
| DY107729.1 | - | 1 - 2 | 235 | 101 | ||
More Information
Set Membership: |
|

WDR20
Genomic Map