Assay Details
| Gene Symbol: CNNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191172.1 | NP_001178101.1 | 4 - 5 | 2070 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNNM2| Gene Symbol: CNNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191172.1 | NP_001178101.1 | 4 - 5 | 2070 | 74 | |
Set Membership: |
|