Assay Details
| Gene Symbol: CTTNBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191181.2 | NP_001178110.1 | 3 - 4 | 464 | 68 | |
| GenBank mRNA | BC111189.1 | - | 6 - 7 | 629 | 68 | |
More Information
Set Membership: |
|

CTTNBP2NL
Genomic Map