Assay Details
| Gene Symbol: ZBTB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191247.3 | NP_001178176.1 | 2 - 3 | 1510 | 64 | |
| GenBank mRNA | DN283168.1 | - | 1 - 2 | 527 | 64 | |
More Information
Set Membership: |
|

ZBTB7A
Genomic Map