Assay Details
| Gene Symbol: GBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191277.2 | NP_001178206.2 | 3 - 4 | 1103 | 113 | |
| GenBank mRNA | DV782751.1 | - | 1 - 2 | 96 | 113 | |
| GH312588.1 | - | 1 - 2 | 550 | 113 | ||
More Information
Set Membership: |
|

GBA3
Genomic Map