Assay Details
| Gene Symbol: GBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191277.2 | NP_001178206.2 | 4 - 5 | 1225 | 88 | |
| GenBank mRNA | DT855153.1 | - | 2 - 3 | 278 | 88 | |
| DV782751.1 | - | 2 - 3 | 218 | 88 | ||
More Information
Set Membership: |
|

GBA3
Genomic Map