Assay Details
| Gene Symbol: LTB4R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191287.1 | NP_001178216.1 | 1 - 2 | 154 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191287.1 transcripts.
Set Membership: |
|

LTB4R2
Genomic Map