Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206812.2 | NP_001193741.1 | 2 - 3 | 601 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZKSCAN7| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206812.2 | NP_001193741.1 | 2 - 3 | 601 | 99 | |
Set Membership: |
|