Assay Details
| Gene Symbol: IER5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206835.1 | NP_001193764.1 | 1 | 1420 | 89 | |
| GenBank mRNA | CO871776.1 | - | 1 | 388 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IER5L| Gene Symbol: IER5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206835.1 | NP_001193764.1 | 1 | 1420 | 89 | |
| GenBank mRNA | CO871776.1 | - | 1 | 388 | 89 | |
Set Membership: |
|