Assay Details
| Gene Symbol: CUTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206896.1 | NP_001193825.1 | 5 - 6 | 573 | 57 | |
| GenBank mRNA | BC109689.1 | - | 5 - 6 | 344 | 57 | |
| EH377683.1 | - | 5 - 6 | 593 | 57 | ||
More Information
Set Membership: |
|

CUTC
Genomic Map