Assay Details
| Gene Symbol: SLC6A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206903.1 | NP_001193832.1 | 10 - 11 | 1407 | 65 | |
| GenBank mRNA | DY459229.1 | - | 3 - 4 | 384 | 65 | |
More Information
Set Membership: |
|

SLC6A11
Genomic Map