Assay Details
| Gene Symbol: ABCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206909.1 | NP_001193838.1 | 20 - 21 | 2458 | 61 | |
| GenBank mRNA | EE344169.1 | - | 5 - 6 | 556 | 61 | |
More Information
Set Membership: |
|

ABCF1
Genomic Map