Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206930.1 | NP_001193859.1 | 23 - 24 | 3743 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUT4| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206930.1 | NP_001193859.1 | 23 - 24 | 3743 | 110 | |
Set Membership: |
|