Assay Details
| Gene Symbol: AOPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206980.2 | NP_001193909.2 | 6 - 7 | 1713 | 66 | |
| GenBank mRNA | BE684589.1 | - | 2 - 3 | 256 | 66 | |
More Information
Set Membership: |
|

AOPEP
Genomic Map