Assay Details
| Gene Symbol: AOPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206980.2 | NP_001193909.2 | 8 - 9 | 1919 | 64 | |
| GenBank mRNA | BE684589.1 | - | 4 - 5 | 462 | 64 | |
| DV916833.1 | - | 2 - 3 | 256 | 64 | ||
More Information
Set Membership: |
|

AOPEP
Genomic Map