Assay Details
| Gene Symbol: CDC20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206982.1 | NP_001193911.1 | 11 - 12 | 1616 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC20B| Gene Symbol: CDC20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206982.1 | NP_001193911.1 | 11 - 12 | 1616 | 59 | |
Set Membership: |
|