Assay Details
| Gene Symbol: CALM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242572.1 | NP_001229501.1 | 5 - 6 | 578 | 78 | |
| GenBank mRNA | BC123890.1 | - | 5 - 6 | 578 | 78 | |
More Information
Set Membership: |
|

CALM1
Genomic Map