Assay Details
Gene Symbol: SYNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243287.1 | NP_001230216.1 | 5 - 6 | 885 | 72 | |
GenBank mRNA | BC149244.1 | - | 5 - 6 | 886 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
