Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243320.1 | NP_001230249.1 | 19 - 20 | 2887 | 122 | |
| GenBank mRNA | BC148910.1 | - | 19 - 20 | 2854 | 122 | |
| EE366789.1 | - | 2 - 3 | 267 | 122 | ||
More Information
Set Membership: |
|

EPRS1
Genomic Map