Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243320.1 | NP_001230249.1 | 22 - 23 | 3418 | 92 | |
| GenBank mRNA | EE366789.1 | - | 5 - 6 | 798 | 92 | |
| EE377545.1 | - | 1 - 2 | 113 | 92 | ||
More Information
Set Membership: |
|

EPRS1
Genomic Map