Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243320.1 | NP_001230249.1 | 27 - 28 | 4022 | 75 | |
| GenBank mRNA | EE377545.1 | - | 6 - 7 | 717 | 75 | |
More Information
Set Membership: |
|

EPRS1
Genomic Map