Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243320.1 | NP_001230249.1 | 2 - 3 | 247 | 98 | |
| GenBank mRNA | BC148910.1 | - | 2 - 3 | 213 | 98 | |
| EE358895.1 | - | 2 - 3 | 247 | 98 | ||
More Information
Set Membership: |
|

EPRS1
Genomic Map