Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243320.1 | NP_001230249.1 | 4 - 5 | 508 | 87 | |
| GenBank mRNA | BC148910.1 | - | 4 - 5 | 474 | 87 | |
| EE358895.1 | - | 4 - 5 | 508 | 87 | ||
More Information
Set Membership: |
|

EPRS1
Genomic Map