Assay Details
| Gene Symbol: SMIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244119.1 | NP_001231048.1 | 1 - 2 | 243 | 62 | |
| GenBank mRNA | BC103022.1 | - | 1 - 2 | 243 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103022.1, NM_001244119.1 transcripts.
Set Membership: |
|

SMIM3
Genomic Map