Assay Details
| Gene Symbol: AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244127.1 | NP_001231056.1 | 5 - 6 | 2231 | 58 | |
| GenBank mRNA | AY862875.1 | - | 2 - 3 | 437 | 58 | |
More Information
Set Membership: |
|

AR
Genomic Map