Assay Details
| Gene Symbol: NEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244145.1 | NP_001231074.1 | 1251 | 85 | ||
| GenBank mRNA | BC151624.1 | - | 2 - 3 | 1242 | 85 | |
More Information
Set Membership: |
|

NEFM
Genomic Map