Assay Details
| Gene Symbol: MCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244177.1 | NP_001231106.1 | 1 - 2 | 464 | 61 | |
| GenBank mRNA | EH153606.1 | - | 1 - 2 | 465 | 61 | |
More Information
Set Membership: |
|

MCAT
Genomic Map